CAPS, 99%, for biochemistry
CAPS, 99%, for biochemistry
Thermo Scientific Chemicals

CAPS, 99%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 172621000
Price (USD)/ Each
76.65
Online exclusive
85.20 
Save 8.55 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
76.65
Online exclusive
85.20 
Save 8.55 (10%)
Add to cart
CAPS, 99%, for biochemistry
Catalog number172621000
Price (USD)/ Each
76.65
Online exclusive
85.20 
Save 8.55 (10%)
-
Add to cart
Chemical Identifiers
CAS1135-40-6
IUPAC Name3-(cyclohexylamino)propane-1-sulfonic acid
Molecular FormulaC9H19NO3S
InChI KeyPJWWRFATQTVXHA-UHFFFAOYSA-N
SMILESOS(=O)(=O)CCCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV(0.1 M in water)
Appearance (Color)White
Titration with NaOH>=98.5 % (On dry substance)
UVat 260 nm A: =<0.04
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only