Thermo Scientific Chemicals

MOPS, 99%, for biochemistry

Catalog number: 172631000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

MOPS, 99%, for biochemistry

Catalog number: 172631000
100 g, Plastic bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 172631000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1132-61-2
IUPAC Name
4-(3-sulfonatopropyl)morpholin-4-ium
Molecular Formula
C7H15NO4S
InChI Key
DVLFYONBTKHTER-UHFFFAOYSA-N
SMILES
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
Appearance (Form)
Crystalline powder
Titration with NaOH
>=98.5 %
UV
at 260 nm A: =<0.05
pH
3.0 to 4.5 (1 % in water at 25°C)
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text