Thermo Scientific Chemicals

Tricine, 98%, for biochemistry

Catalog number: 172641000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tricine, 98%, for biochemistry

Catalog number: 172641000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 172641000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5704-04-1
IUPAC Name
2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}acetic acid
Molecular Formula
C6H13NO5
InChI Key
SEQKRHFRPICQDD-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)NCC(O)=O
Appearance (Form)
Powder
Titration with HClO4
>=97.5 % (On dried basis)
pH
4.0 to 6.0 (1 % in water at 20°C)
UV
at 260 nm A: =<0.04
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text