Thermo Scientific Chemicals

Bicine, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 172652500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Bicine, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 172652500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172652500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
150-25-4
IUPAC Name
sodium 2-[bis(2-hydroxyethyl)amino]acetate
Molecular Formula
C6H12NNaO4
InChI Key
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
SMILES
[Na+].OCCN(CCO)CC([O-])=O
Appearance (Color)
White
Titration with HClO4
>=99.0 %
Sulfated ash
=<0.1 %
Sulfate (SO4)
=<50 ppm
Heavy metals (as Pb)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text