Bis(dimethylamino)phosphorochloridate, 90%, Tech., Thermo Scientific Chemicals
Bis(dimethylamino)phosphorochloridate, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(dimethylamino)phosphorochloridate, 90%, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 172720500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1605-65-8
IUPAC Namebis(dimethylamino)phosphinoyl chloride
Molecular FormulaC4H12ClN2OP
InChI KeyWYLQARGYFXBZMD-UHFFFAOYSA-N
SMILESCN(C)P(Cl)(=O)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4650 to 1.4680 (20°C, 589 nm)
Infrared spectrumConforms
Titration Argentometric88 to 112 % (ex Cl)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only