Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D, Thermo Scientific Chemicals
Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
20 g
Packaging:
Plastic Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 172890200
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.20 
Save 10.55 (15%)
-
Add to cart
Quantity:
20 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.20 
Save 10.55 (15%)
Add to cart
Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D, Thermo Scientific Chemicals
Catalog number172890200
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.20 
Save 10.55 (15%)
-
Add to cart
Chemical Identifiers
CAS14014-06-3
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration29 to 32 % w/w
Appearance (Form)Solution
Appearance (Color)Clear colorless
D-Enrichment>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only