Trimethyl orthobenzoate, 97%, Thermo Scientific Chemicals
Trimethyl orthobenzoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethyl orthobenzoate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 172920500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS707-07-3
IUPAC Name(trimethoxymethyl)benzene
Molecular FormulaC10H14O3
InChI KeyIECKAVQTURBPON-UHFFFAOYSA-N
SMILESCOC(OC)(OC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4870 to 1.4910 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only