Thermo Scientific Chemicals

Triphenylcarbenium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog number: 172950050
5 g, Each
Thermo Scientific Chemicals

Triphenylcarbenium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog number: 172950050
5 g, Each
Quantity
Catalog number: 172950050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
341-02-6
IUPAC Name
tetrafluoroboranuide; triphenylmethylium
Molecular Formula
C19H15BF4
InChI Key
VQXBOEYKSVVPSP-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1
Melting point
203°C to 213°C (Dec)
Appearance (Color)
Yellow to green to brown
Infrared spectrum
Conforms
NMR
>=96.0 %
Appearance (Form)
Powder or crystals and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text