3-Aminobenzonitrile, 99%, Thermo Scientific Chemicals
3-Aminobenzonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminobenzonitrile, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 173040050
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.00 
Save 4.35 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.00 
Save 4.35 (14%)
Add to cart
3-Aminobenzonitrile, 99%, Thermo Scientific Chemicals
Catalog number173040050
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.00 
Save 4.35 (14%)
-
Add to cart
Chemical Identifiers
CAS2237-30-1
IUPAC Name3-aminobenzonitrile
Molecular FormulaC7H6N2
InChI KeyNJXPYZHXZZCTNI-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Infrared spectrumConforms
Appearance (Form)Crystalline solid
Titration with HClO4>=98.5 %
Melting point50°C to 54°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only