Sodium methoxide, pure, anhydrous powder, Thermo Scientific Chemicals
Sodium methoxide, pure, anhydrous powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium methoxide, pure, anhydrous powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
100 g
500 g
Packaging:
View all
Plastic Drum
Catalog number 173120025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Alkalinity>=99 % (Total)
Appearance (Color)White to yellow
Titration>=97 %
NaOH + Na2CO3 (as NaOH)=<2 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only