Thermo Scientific Chemicals

Abietic acid, 85%, Thermo Scientific Chemicals

Catalog number: 173131000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Abietic acid, 85%, Thermo Scientific Chemicals

Catalog number: 173131000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 173131000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
514-10-3
IUPAC Name
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
Molecular Formula
C20H30O2
InChI Key
RSWGJHLUYNHPMX-ONCXSQPRSA-N
SMILES
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
GC
>=80 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text