Thermo Scientific Chemicals

Adipoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 173185000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Adipoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 173185000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 173185000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
111-50-2
IUPAC Name
hexanedioyl dichloride
Molecular Formula
C6H8Cl2O2
InChI Key
PWAXUOGZOSVGBO-UHFFFAOYSA-N
SMILES
ClC(=O)CCCCC(Cl)=O
Appearance (Form)
Liquid
Titration RCOOH
=<2 % (as Adipic acid)
Appearance (Color)
Clear brown
Infrared spectrum
Conforms
Titration RCOCl
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text