Thermo Scientific Chemicals

Fluorescein, disodium salt, pure

Catalog number: 173245000
500 g, Glass Bottle, Each
Thermo Scientific Chemicals

Fluorescein, disodium salt, pure

Catalog number: 173245000
500 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 173245000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
518-47-8
IUPAC Name
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Formula
C20H10Na2O5
InChI Key
NJDNXYGOVLYJHP-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Appearance (Form)
Powder
UV Lambda max
488 to 491 nm (in buffer pH 8)
Appearance (Color)
Orange-red to green-brown to dark brown
Infrared spectrum
Conforms
Loss on drying
=<10 % (105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text