Thermo Scientific Chemicals

2-Hydroxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: 173330010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydroxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: 173330010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 173330010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-01-7
IUPAC Name
2-(hydroxymethyl)phenol
Molecular Formula
C7H8O2
InChI Key
CQRYARSYNCAZFO-UHFFFAOYSA-N
SMILES
OCC1=CC=CC=C1O
Appearance (Form)
Powder or flakes
Appearance (Color)
Light beige
Infrared spectrum
Conforms
GC
>=96.0 %
Melting point
83°C to 87°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text