3-Methylsalicylic acid, 99%, Thermo Scientific Chemicals
3-Methylsalicylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylsalicylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 173341000
Price (USD)/ Each
32.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
38.20 
Save 5.55 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
32.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
38.20 
Save 5.55 (15%)
Add to cart
3-Methylsalicylic acid, 99%, Thermo Scientific Chemicals
Catalog number173341000
Price (USD)/ Each
32.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
38.20 
Save 5.55 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-hydroxy-3-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyWHSXTWFYRGOBGO-UHFFFAOYSA-N
SMILESCC1=CC=CC(C(O)=O)=C1O
Molecular Weight (g/mol)152.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<0.3 % (K.F.)
Appearance (Color)White to off-white to light pink
Melting point163°C to 167°C
Titration with NaOH98.5 to 101.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only