Ethyl 4-bromobutyrate, 97+%, Thermo Scientific Chemicals
Ethyl 4-bromobutyrate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-bromobutyrate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 173350025
Price (USD)/ Each
943.65
Online exclusive
1048.00 
Save 104.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
943.65
Online exclusive
1048.00 
Save 104.35 (10%)
Add to cart
Ethyl 4-bromobutyrate, 97+%, Thermo Scientific Chemicals
Catalog number173350025
Price (USD)/ Each
943.65
Online exclusive
1048.00 
Save 104.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2969-81-5
IUPAC Nameethyl 4-bromobutanoate
Molecular FormulaC6H11BrO2
InChI KeyXBPOBCXHALHJFP-UHFFFAOYSA-N
SMILESCCOC(=O)CCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.0 %
Appearance (Form)Liquid
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only