Ethyl 4-bromobutyrate, 97+%, Thermo Scientific Chemicals
Ethyl 4-bromobutyrate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-bromobutyrate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number 173355000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameethyl 4-bromobutanoate
Molecular FormulaC6H11BrO2
InChI KeyXBPOBCXHALHJFP-UHFFFAOYSA-N
SMILESCCOC(=O)CCCBr
Molecular Weight (g/mol)195.06
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.0 %
Appearance (Form)Liquid
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only