Methyl DL-2-bromopropionate, 99%, Thermo Scientific Chemicals
Methyl DL-2-bromopropionate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl DL-2-bromopropionate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 173391000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5445-17-0
IUPAC Namemethyl 2-bromopropanoate
Molecular FormulaC4H7BrO2
InChI KeyACEONLNNWKIPTM-UHFFFAOYNA-N
SMILESCOC(=O)C(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4500 to 1.4530 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only