Chemical Identifiers
IUPAC Namedisodium 2-hydroxy-5-[4,5,6,7-tetrabromo-1-(4-hydroxy-3-sulfonatophenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]benzene-1-sulfonate
Molecular FormulaC20H8Br4Na2O10S2
InChI KeyGHAFORRTMVIXHS-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O
Molecular Weight (g/mol)837.99
View more