Thermo Scientific Chemicals

1-Iodobutane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 173435000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

1-Iodobutane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 173435000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 173435000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
542-69-8
IUPAC Name
1-iodobutane
Molecular Formula
C4H9I
InChI Key
KMGBZBJJOKUPIA-UHFFFAOYSA-N
SMILES
CCCCI
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Clear colorless to light yellow to light pink
Infrared spectrum
Conforms
Refractive index
1.4975 to 1.4995 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text