ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Glass bottle
Catalog number 173440010
Price (USD)/ Each
132.65
Online exclusive
147.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
132.65
Online exclusive
147.00 
Save 14.35 (10%)
Add to cart
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Catalog number173440010
Price (USD)/ Each
132.65
Online exclusive
147.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS502-44-3
IUPAC Nameoxepan-2-one
Molecular FormulaC6H10O2
InChI KeyPAPBSGBWRJIAAV-UHFFFAOYSA-N
SMILESO=C1CCCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4620 to 1.4640 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only