Thermo Scientific Chemicals

tert-Butyl carbamate, 98%, Thermo Scientific Chemicals

Catalog number: 173450250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

tert-Butyl carbamate, 98%, Thermo Scientific Chemicals

Catalog number: 173450250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 173450250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4248-19-5
IUPAC Name
tert-butyl carbamate
Molecular Formula
C5H11NO2
InChI Key
LFKDJXLFVYVEFG-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(N)=O
Appearance (Color)
White to light yellow
Appearance (Form)
Powder or crystals or needles
Melting point
106°C to 110°C
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text