Thermo Scientific Chemicals

Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals

Catalog number: 173460100
10 g, Glass Bottle, Each
Thermo Scientific Chemicals

Celite™, for analysis, high purity analytical grade, Thermo Scientific Chemicals

Catalog number: 173460100
10 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 173460100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61790-53-2
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Appearance (Form)
Powder
Conductivity
=<15 µS/cm
Additional info
SiO2: 98.7%
Additional info
Nonsiliceous substances < 50 mg
pH
5 to 8 (25°C, 10% aqueous suspension)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text