Thermo Scientific Chemicals

Chloroacetone, 96%, stabilized, Thermo Scientific Chemicals

Catalog number: 173470050
5 L, Plastic drum, Each
Thermo Scientific Chemicals

Chloroacetone, 96%, stabilized, Thermo Scientific Chemicals

Catalog number: 173470050
5 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 173470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
78-95-5
IUPAC Name
1-chloropropan-2-one
Molecular Formula
C3H5ClO
InChI Key
BULLHNJGPPOUOX-UHFFFAOYSA-N
SMILES
CC(=O)CCl
Appearance (Color)
Clear yellow to yellow-brown
Infrared spectrum
Conforms
Refractive index
1.4310 to 1.4330 (20°C, 589 nm)
GC
>=95 %
Stabilizer
0.1 % epoxidized soybean oil

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text