Chloroacetone, 96%, stabilized, Thermo Scientific Chemicals
Chloroacetone, 96%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroacetone, 96%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 173472500
Price (USD)/ Each
46.65
Online exclusive
52.20 
Save 5.55 (11%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
52.20 
Save 5.55 (11%)
Add to cart
Chloroacetone, 96%, stabilized, Thermo Scientific Chemicals
Catalog number173472500
Price (USD)/ Each
46.65
Online exclusive
52.20 
Save 5.55 (11%)
-
Add to cart
Chemical Identifiers
CAS78-95-5
IUPAC Name1-chloropropan-2-one
Molecular FormulaC3H5ClO
InChI KeyBULLHNJGPPOUOX-UHFFFAOYSA-N
SMILESCC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to yellow-brown
Infrared spectrumConforms
Refractive index1.4310 to 1.4330 (20°C, 589 nm)
Stabilizer0.1 % epoxidized soybean oil
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only