4-Bromo-2-chlorophenol, 99%
4-Bromo-2-chlorophenol, 99%
4-Bromo-2-chlorophenol, 99%
4-Bromo-2-chlorophenol, 99%
Thermo Scientific Chemicals

4-Bromo-2-chlorophenol, 99%

CAS: 3964-56-5 | C6H4BrClO | 207.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 173530050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3964-56-5
IUPAC Name4-bromo-2-chlorophenol
Molecular FormulaC6H4BrClO
InChI KeyVIBJPUXLAKVICD-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles and/or chunks or
Melting point47°C to 51°C
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)low melting mass
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link