Thermo Scientific Chemicals

4-Bromo-2-chlorophenol, 99%, Thermo Scientific Chemicals

Catalog number: 173530250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromo-2-chlorophenol, 99%, Thermo Scientific Chemicals

Catalog number: 173530250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 173530250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3964-56-5
IUPAC Name
4-bromo-2-chlorophenol
Molecular Formula
C6H4BrClO
InChI Key
VIBJPUXLAKVICD-UHFFFAOYSA-N
SMILES
OC1=CC=C(Br)C=C1Cl
Appearance (Form)
Crystalline powder or needles and/or chunks or
Melting point
47°C to 51°C
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Appearance (Form)
low melting mass

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text