Ethyl 2-oxocyclopentanecarboxylate, 95+%, Thermo Scientific Chemicals
Ethyl 2-oxocyclopentanecarboxylate, 95+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-oxocyclopentanecarboxylate, 95+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 173580250
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.50 
Save 6.85 (14%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.50 
Save 6.85 (14%)
Add to cart
Ethyl 2-oxocyclopentanecarboxylate, 95+%, Thermo Scientific Chemicals
Catalog number173580250
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.50 
Save 6.85 (14%)
-
Add to cart
Chemical Identifiers
CAS611-10-9
IUPAC Nameethyl 2-oxocyclopentane-1-carboxylate
Molecular FormulaC8H12O3
InChI KeyJHZPNBKZPAWCJD-UHFFFAOYNA-N
SMILESCCOC(=O)C1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=95.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4500 to 1.4540 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only