Thermo Scientific Chemicals

4,4'-Biphenol, 97%, Thermo Scientific Chemicals

Catalog number: 173672500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

4,4'-Biphenol, 97%, Thermo Scientific Chemicals

Catalog number: 173672500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 173672500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92-88-6
IUPAC Name
[1,1'-biphenyl]-4,4'-diol
Molecular Formula
C12H10O2
InChI Key
VCCBEIPGXKNHFW-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1=CC=C(O)C=C1
Appearance (Color)
Off-white to light beige
Infrared spectrum
Conforms
Appearance (Form)
Powder
GC
>=96.0 %
Melting point
278°C to 283°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text