Thermo Scientific Chemicals

Neopentylamine, 97%, Thermo Scientific Chemicals

Catalog number: 173691000
100 mL, Each
Thermo Scientific Chemicals

Neopentylamine, 97%, Thermo Scientific Chemicals

Catalog number: 173691000
100 mL, Each
Quantity
Catalog number: 173691000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5813-64-9
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4010 to 1.4050 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text