1-Docosanol, 98%, Thermo Scientific Chemicals
1-Docosanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Docosanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 173721000
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
Add to cart
1-Docosanol, 98%, Thermo Scientific Chemicals
Catalog number173721000
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS661-19-8
IUPAC Namedocosan-1-ol
Molecular FormulaC22H46O
InChI KeyNOPFSRXAKWQILS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Melting point68°C to 72°C
Appearance (Form)Pellets or tablets
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only