L(+)-Lysine monohydrate, 99%
L(+)-Lysine monohydrate, 99%
L(+)-Lysine monohydrate, 99%
L(+)-Lysine monohydrate, 99%
Thermo Scientific Chemicals

L(+)-Lysine monohydrate, 99%

CAS: 39665-12-8 | C6H16N2O3 | 164.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
Catalog number 173730250
Price (EUR)
46,16
Online Exclusive
54,30
Save 8,14 (15%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Plastic Bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
46,16
Online Exclusive
54,30
Save 8,14 (15%)
Each
Add to cart
Chemical Identifiers
CAS39665-12-8
IUPAC Name(2S)-2,6-diaminohexanoic acid hydrate
Molecular FormulaC6H16N2O3
InChI KeyHZRUTVAFDWTKGD-XZNNNFJINA-N
SMILESO.NCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Other amino acids=<0.5 % (TLC)
Specific optical rotation(On dry basis)
Appearance (Form)Crystalline powder or crystals
Titration with HClO498.5 to 101.5 %
Sulfated ash=<0.1 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471823Certificate of AnalysisMay 17, 2025173730250, 173735000, 173731000
A0468496Certificate of AnalysisJan 31, 2025173730250, 173735000, 173731000
A0461687Certificate of AnalysisApr 18, 2024173730250, 173735000, 173731000
A0449068Certificate of AnalysisAug 25, 2023173730250, 173735000, 173731000
A0435217Certificate of AnalysisMay 02, 2023173730250, 173731000, 173735000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection