Thermo Scientific Chemicals

L(+)-Lysine monohydrate, 99%

Catalog number: 173735000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

L(+)-Lysine monohydrate, 99%

Catalog number: 173735000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 173735000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39665-12-8
IUPAC Name
(2S)-2,6-diaminohexanoic acid hydrate
Molecular Formula
C6H16N2O3
InChI Key
HZRUTVAFDWTKGD-XZNNNFJINA-N
SMILES
O.NCCCC[C@H](N)C(O)=O
Other amino acids
=<0.5 % (TLC)
Specific optical rotation
(On dry basis)
Appearance (Form)
Crystalline powder or crystals
Titration with HClO4
98.5 to 101.5 %
Sulfated ash
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text