Thermo Scientific Chemicals

Pyruvic aldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals

Catalog number: 173805000
500 g, Each
Thermo Scientific Chemicals

Pyruvic aldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals

Catalog number: 173805000
500 g, Each
Quantity
Catalog number: 173805000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6342-56-9
IUPAC Name
1,1-dimethoxypropan-2-one
Molecular Formula
C5H10O3
InChI Key
ULVSHNOGEVXRDR-UHFFFAOYSA-N
SMILES
COC(OC)C(C)=O
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow
Refractive index
1.3970 to 1.3990 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text