Thermo Scientific Chemicals

α-D-Methylglucoside, 99%, Thermo Scientific Chemicals

Catalog number: 173841000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

α-D-Methylglucoside, 99%, Thermo Scientific Chemicals

Catalog number: 173841000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 173841000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-30-3
IUPAC Name
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular Formula
C7H14O6
InChI Key
HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES
COC1OC(CO)C(O)C(O)C1O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Loss on drying
=<0.1 % (60 to 70°C)
Specific optical rotation
+156° to +161° (20°C, 589 nm) (c=10, water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text