5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals
5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
50 g
Packaging:
Glass bottle
Catalog number 173860500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS698-63-5
IUPAC Name5-nitrofuran-2-carbaldehyde
Molecular FormulaC5H3NO4
InChI KeySXINBFXPADXIEY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(O1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Low melting crystalline solid
Melting point33°C to 37°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only