n-Amylamine, 99%, Thermo Scientific Chemicals
n-Amylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Amylamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
5 mL
250 mL
Packaging:
Glass bottle
Catalog number 173940500
Price (USD)/ Each
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-58-7
IUPAC Namepentan-1-amine
Molecular FormulaC5H13N
InChI KeyDPBLXKKOBLCELK-UHFFFAOYSA-N
SMILESCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
AppearanceMay darken on storage
Water=<1 %
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only