Thermo Scientific Chemicals

Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals

Catalog number: 173961000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Pentadecafluorooctanoic acid, 96%, Thermo Scientific Chemicals

Catalog number: 173961000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 173961000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
335-67-1
IUPAC Name
pentadecafluorooctanoic acid
Molecular Formula
C8HF15O2
InChI Key
SNGREZUHAYWORS-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Water
=<4.5 % (K.F.)
Titration with NaOH
>=95 %
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text