Thermo Scientific Chemicals

Hydrocinnamonitrile, 99+%, Thermo Scientific Chemicals

Catalog number: 173990100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Hydrocinnamonitrile, 99+%, Thermo Scientific Chemicals

Catalog number: 173990100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 173990100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
645-59-0
IUPAC Name
3-phenylpropanenitrile
Molecular Formula
C9H9N
InChI Key
ACRWYXSKEHUQDB-UHFFFAOYSA-N
SMILES
N#CCCC1=CC=CC=C1
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.5210 to 1.5230 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=99.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text