Thermo Scientific Chemicals

1,2-Dicyanobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 174012500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

1,2-Dicyanobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 174012500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 174012500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
91-15-6
IUPAC Name
benzene-1,2-dicarbonitrile
Molecular Formula
C8H4N2
InChI Key
XQZYPMVTSDWCCE-UHFFFAOYSA-N
SMILES
N#CC1=CC=CC=C1C#N
Melting point
138°C to 142°C
Appearance (Color)
White to beige to grey-green
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text