DL-Proline, 99%
Thermo Scientific Chemicals

DL-Proline, 99%

Have Questions?
Quantity:
25 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 174030250
Price (USD)/ Each
117.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
138.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
117.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
138.00 
Save 20.35 (15%)
Add to cart
DL-Proline, 99%
Catalog number174030250
Price (USD)/ Each
117.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
138.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepyrrolidine-2-carboxylic acid
Molecular FormulaC5H9NO2
InChI KeyONIBWKKTOPOVIA-UHFFFAOYNA-N
SMILESOC(=O)C1CCCN1
Molecular Weight (g/mol)115.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Titration with HClO498.5 to 101.5 %
Infrared spectrumConforms
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only