L(+)-Rhamnose monohydrate, 99%
L(+)-Rhamnose monohydrate, 99%
L(+)-Rhamnose monohydrate, 99%
L(+)-Rhamnose monohydrate, 99%
Thermo Scientific Chemicals

L(+)-Rhamnose monohydrate, 99%

CAS: 10030-85-0 | C6H12O5 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 174085000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10030-85-0
IUPAC Name(2R,3R,4R,5R,6S)-6-methyloxane-2,3,4,5-tetrol
Molecular FormulaC6H12O5
InChI KeySHZGCJCMOBCMKK-HGVZOGFYSA-N
SMILESC[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Heavy metals=<20 ppm
Water9.0 to 11.0 % (K.F.)
Residue after ignition=<0.2 %
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link