Tetrabutylammonium chloride, 95%, Tech., contains also bromide
Tetrabutylammonium chloride, 95%, Tech., contains also bromide
Tetrabutylammonium chloride, 95%, Tech., contains also bromide
Tetrabutylammonium chloride, 95%, Tech., contains also bromide
Thermo Scientific Chemicals

Tetrabutylammonium chloride, 95%, Tech., contains also bromide

CAS: 1112-67-0 | C16H36ClN | 277.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 174120250
Price (USD)
83.65
Online Exclusive
92.50
Save 8.85 (10%)
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
92.50
Save 8.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS1112-67-0
IUPAC Nametetrabutylazanium chloride
Molecular FormulaC16H36ClN
InChI KeyNHGXDBSUJJNIRV-UHFFFAOYSA-M
SMILES[Cl-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)chunks or fused solid
Titration Argentometric>=94 % (On dry substance)
Bromide (Br)=<5 %
Appearance (Form)Crystalline powder or crystals or
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468270Certificate of AnalysisJan 31, 2025174120050, 174121000, 174120250, 174125000
A0463282Certificate of AnalysisJun 12, 2024174120050, 174121000, 174120250, 174125000
A0409195Certificate of AnalysisDec 13, 2022174120050, 174121000, 174120250, 174125000
A0446648Certificate of AnalysisOct 25, 2022174120050, 174121000, 174120250, 174125000
A0440006Certificate of AnalysisMay 14, 2022174120050, 174121000, 174120250, 174125000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection