Tetradecane, 99%, Thermo Scientific Chemicals
Tetradecane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetradecane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
500 mL
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 174130025
Price (USD)/ Each
636.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
749.00 
Save 112.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
636.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
749.00 
Save 112.35 (15%)
Add to cart
Tetradecane, 99%, Thermo Scientific Chemicals
Catalog number174130025
Price (USD)/ Each
636.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
749.00 
Save 112.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametetradecane
Molecular FormulaC14H30
InChI KeyBGHCVCJVXZWKCC-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC
Molecular Weight (g/mol)198.39
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.4280 to 1.4300 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only