Thermo Scientific Chemicals

Tetradecane, 99%, Thermo Scientific Chemicals

Catalog number: 174131000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Tetradecane, 99%, Thermo Scientific Chemicals

Catalog number: 174131000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 174131000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
629-59-4
IUPAC Name
tetradecane
Molecular Formula
C14H30
InChI Key
BGHCVCJVXZWKCC-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCC
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Refractive index
1.4280 to 1.4300 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text