Thermo Scientific Chemicals

Tiron, Thermo Scientific Chemicals

Catalog number: 174145000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Tiron, Thermo Scientific Chemicals

Catalog number: 174145000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 174145000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
149-45-1
IUPAC Name
disodium 4,5-dihydroxybenzene-1,3-disulfonate
Molecular Formula
C6H4Na2O8S2
InChI Key
ISWQCIVKKSOKNN-UHFFFAOYSA-L
SMILES
[Na+].[Na+].OC1=CC(=CC(=C1O)S([O-])(=O)=O)S([O-])(=O)=O
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Appearance (Form)
Powder
Titration with NaOH
>=98.0 %
Water
4.5 to 6.0 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text