DL-2-Amino-4-pentenoic acid, 99%, Thermo Scientific Chemicals
DL-2-Amino-4-pentenoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-2-Amino-4-pentenoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 174220050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7685-44-1
IUPAC Name(2R)-2-azaniumylpent-4-enoate
Molecular FormulaC5H9NO2
InChI KeyWNNNWFKQCKFSDK-SCSAIBSYSA-N
SMILES[NH3+][C@H](CC=C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=98.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only