Thermo Scientific Chemicals

Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals

Catalog number: 174281000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals

Catalog number: 174281000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 174281000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
940-71-6
IUPAC Name
hexachloro-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Molecular Formula
Cl6N3P3
InChI Key
UBIJTWDKTYCPMQ-UHFFFAOYSA-N
SMILES
ClP1(Cl)=NP(Cl)(Cl)=NP(Cl)(Cl)=N1
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Crystalline powder
Melting point
111°C to 115°C
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text