2-Methoxypropene, 97%, stabilized, Thermo Scientific Chemicals
2-Methoxypropene, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxypropene, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
Packaging:
Glass bottle
Catalog number 174350010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-methoxyprop-1-ene
Molecular FormulaC4H8O
InChI KeyYOWQWFMSQCOSBA-UHFFFAOYSA-N
SMILESCOC(C)=C
Molecular Weight (g/mol)72.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=96.0 %
Stabilizer=<0.5 % potassium carbonate
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only