Thermo Scientific Chemicals

2-Methoxypropene, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 174352500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Methoxypropene, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 174352500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 174352500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
116-11-0
IUPAC Name
2-methoxyprop-1-ene
Molecular Formula
C4H8O
InChI Key
YOWQWFMSQCOSBA-UHFFFAOYSA-N
SMILES
COC(C)=C
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
GC
>=96.0 %
Stabilizer
=<0.5 % potassium carbonate
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text